SP Bel-Art Burettes, Automatic Self-Zeroing, Class B, Bel-Art Products, a part of SP
Proveedor: Bel-Art Products, a Part of SP
Certificados
Acerca de este artículo
These self-zeroing glass burettes have easy-to-read blue graduations against a white background.
- Easy-to-read measurements
- Blue graduations on white background
- Polyethylene construction
- Class B graduation accuracy
The delivery end is fitted into a plastic push button stopcock which permits drop by drop delivery
A polyethylene filling tube extends from the top of the burette into the bottle reservoir. Bottle, screw cap, and round base are polyethylene plastic. Graduation accuracy is Class B.
A small screw clamp adjusts drop speed as needed for titration.
Información de pedido: Borosilicate glass burets come complete with plastic stopcock assembly and polyethylene bottle, filling tube, bottle base, and screw cap.
Frequently Bought Together