Orders LinkContactUsLinkComponent
VWR® Funnels, PTFE
VWR® Funnels, PTFE
VWR® Funnels, PTFE
VWR® Funnels, PTFE
Isostatically molded, FDA approved pure PTFE molding provides a durable, heavy wall construction with a smooth, non-stick internal finish.
  • Chemically inert
  • May be used from 0 to 280 °C (32 to 536 °F)
Suitable for a variety of laboratory applications.
Product Family Options
  • Height
    Stem Diameter
    Top Diameter
  • 50 mm
    8 mm
    30 mm
    Each (1UN)
    Each (1UN) - $850.16
    $850.16
    Each (1UN)
       $850.16
  • 85 mm
    10 mm
    50 mm
    Each (1UN)
    Each (1UN) - $1,155.01
    $1,155.01
    Each (1UN)
       $1,155.01
  • 110 mm
    12 mm
    70 mm
    Each (1UN)
    Each (1UN) - $1,689.63
    $1,689.63
    Each (1UN)
       $1,689.63
  • 170 mm
    14 mm
    100 mm
    Each (1UN)
    Each (1UN) - $3,475.75
    $3,475.75
    Each (1UN)
       $3,475.75