KIMBLE® KONTES® Buchner Funnels, with KIMFLOW® Fritted Disc, Kimble Chase
For use wherever filter paper and membranes cannot withstand the chemical attack of the filtrate.
- Has a marking spot
Funnels are constructed with heavy walled stems and beaded top rims for extra strength.
: Designed from Federal Specification DD-F-776. Low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
: Supplied with KIMFLOW® fritted disc.
Frequently Bought Together