VWR®, Beakers, PTFE
Supplier: VWR International
Jump to:
Acerca de este artículo
Isostatically molded in pure, FDA approved PTFE, these beakers feature a pouring spout and have a machined base for good heat transfer.
- Chemically resistant and inert
- Autoclavable
Machined base provides good heat transfer in these PTFE beakers.
5 Options Available Below
Product Details & Documents
Isostatically molded in pure, FDA approved PTFE, these beakers feature a pouring spout and have a machined base for good heat transfer.
- Chemically resistant and inert
- Autoclavable
Machined base provides good heat transfer in these PTFE beakers.
Documents
Certificates
Buscar por número de loteProduct Family Options
Información del producto
- CapacityHeightExt.ØAvailabilityPrice
Accessories
Información del producto
- Catalog #For Use WithAvailabilityPrice
- Beaker Covers