Orders LinkContactUsLinkComponent
Corning® Spinner Flasks with Vertical Sidearms, Corning
Corning® Spinner Flasks with Vertical Sidearms, Corning
36 L, 120 mm, 45 mm
Catalog # 22877-138
Supplier:   Corning
Número de proveedor #:  4512-36L
Corning® Spinner Flasks with Vertical Sidearms, Corning
Catalog # 22877-138
Supplier:  Corning
Corning® Spinner Flasks with Vertical Sidearms, Corning
36 L, 120 mm, 45 mm
Price
$91,350.00
Log in Para disponibilidad
Case (1UN)
Product Details & Documents
Borosilicate glass flasks are baffled to enhance agitation of contents.
  • Side baffles enhance aeration and agitation of flask contents
  • Proprietary 'stretch' impeller design ensures optimal stirring
  • Vertical sidearms permit use with sidearm probes and fittings
Can be used with direct drive motor or magnetically driven paddle assembly.
Consistent height to width ratios for greater predictability in scale up across the full range of vessel sizes.
Accessories information: Proculture® spinner direct top drive motor with high torque, low rpm specifically designed to maintain constant low speed for use on top of Proculture® spinner flasks. The gearhead stirrer motor delivers 14.5 in-lbs of torque and has a maximum speed of 350 rpm.
100 mm Center Cap Assembly for Proculture® Spinner Flasks is designed for use with direct overhead drive motor, these cap assemblies required optional 8L, 15 L or 36 L paddle assemblies.
Documents
Especificaciones
  • Capacity:
    36 L
  • Sidearm Neck O.D.:
    45 mm
  • Center Neck O.D.:
    120 mm