Nalgene® Large Square Bottle, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Catalog # 16130-010
Supplier: Thermo Fisher Scientific
Specifications
- Capacidad:4 L (1 gal.)
- Referencia:16130-010
- Nº de proveedor:2123-0010
Specifications
About this item
Excellent for collecting, storing, and transporting dry chemicals, environmental samples, reagents, and distilled water.
Useful for low temperature applications. Molded-in handgrips provide greater safety and convenience when pouring. Wide mouth design allows easy access to contents. Leakproof. Cap size: 100-415.
Ordering information: Supplied with polypropylene screw caps assembled on bottles.