ContactUsLinkComponent
Nalgene® Square Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
  16121-004
   Product Transparency and Disclosure
undefined
Nalgene® Square Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
  16121-004
 :  2114-0006
   Product Transparency and Disclosure
  • Tamaño de la tapa:
    38-415
  • Capacidad:
    177 ml (6 oz.)
  • Environmentally Preferable:
    Product Transparency and Disclosure
  • Certificación ambiental:
    ACT Label
  • Referencia:
    16121-004
  • Nº de proveedor:
    2114-0006
Ideal for liquid or chemical storage, these HDPE square bottles make excellent sample mailers.
  • Square shape saves space
  • Bottles can withstand temperatures up to 120 °C (248 °F)
  • Leakproof, Not autoclavable
Wide mouth design allows for easy filling and dispensing.
 : Supplied with polypropylene screw caps assembled on bottles.