Orders LinkContactUsLinkComponent
VWR® Wide Mouth Bottles, Rectangular, HDPE,
VWR® Wide Mouth Bottles, Rectangular, HDPE,
VWR® Wide Mouth Bottles, Rectangular, HDPE,
Supplier:  VWR International
Certificates
VWR® Wide Mouth Bottles, Rectangular, HDPE,
VWR® Wide Mouth Bottles, Rectangular, HDPE,
3 Options Available Below
Product Details & Documents
Space-saving, leakproof bottles are for sampling, storage, or shipment of dry chemicals or liquids.
  • Sturdy construction
  • Space-saving design
  • Leakproof
  • HDPE fabrication
Sturdy construction prevents bulging, translucent material allows for optical clarity, and wide mouth makes filling easy. Bottles have excellent chemical resistance to most acids, bases, and alcohols, and can be stored in freezer to −100 °C (−148 °F). Bottle 414004-195 features integrated handle for better grip, greater safety, and pouring convenience. Polypropylene screw caps included.
Product Family Options
Información del producto
  • Capacity
    Cap Size
    W×L×H
    Availability
    Price
  • 500 ml (16.9 oz.)
    48-415
    64×99.5×114 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 12
    Paquete de 12 - $510.58
    $510.58
    Log in Para disponibilidad
    Case de 4 (12UN)
    Case de 4 (12UN) - $556.53
    $556.53
    Log in Para disponibilidad
    Paleta de 84 (12UN)
    Paleta de 84 (12UN) - $42,888.54
    $42,888.54
    Paquete de 12
       $510.58
    Case de 4 (12UN)
       $556.53
    Paleta de 84 (12UN)
       $42,888.54
  • 1 L (33.8 oz.)
    53-415
    67×127×180 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 6
    Paquete de 6 - $561.09
    $561.09
    Log in Para disponibilidad
    Case de 4 (6UN)
    Case de 4 (6UN) - $611.59
    $611.59
    Log in Para disponibilidad
    Paleta de 84 (6UN)
    Paleta de 84 (6UN) - $47,131.43
    $47,131.43
    Paquete de 6
       $561.09
    Case de 4 (6UN)
       $611.59
    Paleta de 84 (6UN)
       $47,131.43
  • 2 L (67.6 oz.)
    63-415
    96×150×237 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 4
    Paquete de 4 - $618.62
    $618.62
    Log in Para disponibilidad
    Case de 3 (4UN)
    Case de 3 (4UN) - $674.29
    $674.29
    Log in Para disponibilidad
    Paleta de 48 (4UN)
    Paleta de 48 (4UN) - $29,693.69
    $29,693.69
    Paquete de 4
       $618.62
    Case de 3 (4UN)
       $674.29
    Paleta de 48 (4UN)
       $29,693.69