KIMAX® Graduated Cylinders, Class A, To Contain, Serialized and Certified, with Glass Stoppers, Kimble Chase
Cylinders and [ST] stoppers are made of sturdy borosilicate glass
Cylinders are designed from ASTM Specification E1272, Style 2, Class A requirements. Each cylinder is permanently marked with an individual serial number and is supplied with a Certificate of Graduation Accuracy. Stoppers are designed from ASTM Specification E675.
Cylinders are suitable for mixing. Single metric scale is in durable white ceramic enamel.
: Replacement stoppers are available; see the 89002-174 series.
- Catalog No:
- 89001-712
- 89001-716
- 89001-708
- 89001-714
- Forma:
- Short
- Short
- Short
- Short
Frequently Bought Together