KIMAX® Single Metric Scale Graduated Cylinders, Class B, with Polyethylene Stoppers, Kimble Chase
Catálogo # 89000-766
Proveedor: DWK LIFE SCIENCES S. DE R.L. DE MX
Especificaciones
- Intervalo de graduación:1 ml
- Rango de graduación:5–100 mL
- Capacidad:100 ml
- Forma:Tall
- Referencia:89000-766
- [ST] No. de tapón:22
- Height Without Stopper:255 mm
- Tolerancia:±0.6 ml
- Nº de proveedor:20039P-100
Especificaciones
Acerca de este artículo
Calibrated to contain
Cylinders are designed from ASTM Specification E1272, Style 2, Class B requirements.
Cylinders are made of sturdy borosilicate glass. Scale is durable white ceramic enamel. Yellow and blue [ST] stopper is made from linear high-density polyethylene and has a closed bottom. The enlarged top of the stopper protects the neck if the cylinder is knocked over.
Información de pedido: Replacement stoppers are available; see the 89000-797 series.