SP Bel-Art Precisionware™ Bottles, Polypropylene, Wide Mouth, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
For pharmaceutical or similar packaging of liquid or dry materials.
- Polypropylene construction
- Deep, square threads
- Smooth, rounded interiors
- Easy to clean and thoroughly empty
- Autoclavable
Polypropylene bottles are Boston-round style, clear, and molded from FDA compliant material. Polypropylene caps have a valve-seal design for a leakproof fit. Deep, square threads help prevent back-off during shipping.
Ribbed cap exteriors are designed for torque closure and easy opening. Smooth, rounded bottle interiors are designed for easy cleaning and thorough emptying. Autoclavable, with caps detached, to 121 °C (250 °F).
Related Information
Wilmad LabGlass Media Formulation and Transfer for Vaccine Development
Product Literature
Frequently Bought Together