KIMAX® Graduated Cylinders, Class A, To Deliver, Serialized and Certified, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
Cylinders are made of sturdy borosilicate glass with a hexagonal glass base that has been flat ground for stability.
Cylinders feature a single metric scale
Designed from ASTM Specification E1272, Style 1, Class A requirements. Each cylinder is permanently marked with an individual serial number and is supplied with a Certificate of Graduation Accuracy.
The 10mL cylinder has an enlarged funnel-shaped top for ease of filling. Cylinders 25mL and larger are supplied with SAFE-GARD® bumpers, which are sturdy split rings made of yellow low-density polyethylene.
Specifications
- Catalog No:
- 89001-676
- 89001-678
- 89001-672
- 89001-674
- 89001-668
- 89001-670
- 89001-666
- Forma:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
Specifications
Related Information
Frequently Bought Together