ContactUsLinkComponent
Chemware® Teflon® Resin PFA Jars, Saint-Gobain
Catalog # 36324-001
undefined
Chemware® Teflon® Resin PFA Jars, Saint-Gobain
Catalog # 36324-001
Supplier Number:  D1069025
Specifications
  • Capacidad:
    120 ml (4 oz.)
  • Referencia:
    36324-001
  • Nº de proveedor:
    D1069025
Specifications
About this item
Store solutions, pastes, and bulk materials without leaching, corrosion, contamination, or evaporation.
  • Knurled top for sure grip. Large mouth for easy access
  • Secondary inner seal for safe storage of any substance
  • Heat to 260 °C (500 °F). Freeze to –195 °C (–320 °F)
  • Stackable, easily cleaned
Certifications: Jar 36324-007 meets EPA requirement for soil sampling.