SP Bel-Art Wide Mouth Storage Bottles, Heavy-Duty Closure, LDPE, Bel-Art Products, a part of SP
Proveedor: Bel-Art Products, a Part of SP
Certificados
Acerca de este artículo
Low-density polyethylene bottles with a heavy-threaded, self-sealing screw cap closure provide reliability when storing, shipping, and handling materials in both liquid and powder form.
- LDPE construction
- Heavy-threaded, self-sealing screw cap closure
- Reliable for use with both liquid and powder contents
- Larger sizes are made of HDPE
Note: 5000 and 10000 ml sizes are made of high-density polyethylene and have an octagonal shape. They are not leakproof and are best used for dry materials.
Frequently Bought Together