HPLC Poly Manifolds for Quick-Disconnect Safety Disposal Cans, Justrite®
Proveedor: JUSTRITE MANUFACTURING CO
Certificados
Acerca de este artículo
Manifolds enable up to six waste lines (per manifold) from HPLC processes to be fed into a single intake fitting.
- Equipped with six 1/4-28 ports, one 9.5 mm (3/8") NPT port
- Option of one 316 stainless steel quick disconnect, one polypropylene quick disconnect, or 316 stainless steel stacker connector
- Six ports tapped to accommodate 1/4-28 threads of compression fittings
The compression fittings accommodate 1.6 mm (1/16") or 3.2 mm (1/8") O.D tubing sizes. Tefzel® fittings are approximately 2.5 cm (1") long and include ferrules for a compression fit to the waste line.
The manifold with stainless steel stackable connection enables attachment on top of either the stainless steel or polypropylene quick disconnect manifolds. This addition allows up to a total of 12 waste lines to be fed to a disposal can.
Certifications: Safety cans with manifold installed are OSHA compliant and meet NFPA Code 30.
Ordering information: Manifolds include caps for sealing off the unused ports and reducing vapor release. Tubing not included.
Frequently Bought Together