VentSep® Separatory Funnels, Pear-Shape, Glas-Col®
Catalog # 57020-301
Supplier: GLAS-COL
Small Business Enterprise
Specifications
- Capacidad:2000 ml
- Referencia:57020-301
- Nº de proveedor:099ASST655
Specifications
About this item
These separatory funnels feature a built-in relief valve, a fluoropolymer resin stopcock, and a screw cap with fluoropolymer resin liner
Funnels vent gas pressure over 1psi automatically. Funnels are ideal for vigorous hand or machine shaking.