VITLAB® Class B Graduated Cylinders, SAN, BrandTech
83008-912
- Intervalo de graduación:5 ml
- Capacidad:500 ml
- Forma:Short
- Referencia:83008-912
- D.E. x Al:61×350 mm
- Tolerancia:±5 ml
- Nº de proveedor:V65191
These cylinders have ring-molded graduations at major increments to simplify measurements.
- Plastic-clear styrene acrylonitrile copolymer (SAN)
- Molded graduations for durability
These VITLAB® graduated cylinders are made of plastic-clear styrene acrylonitrile copolymer with molded graduations for easy meniscus readings. Hexagonal base resists rolling.
: Class B per DIN 12681.