SP Bel-Art HOLDFAST™ Graduated Cylinders, Polypropylene, Bel-Art Products, a part of SP
24780-371
- Intervalo de graduación:10 ml
- Capacidad:1000 ml
- Altura:17.3"
- Forma:Tall
- Referencia:24780-371
- D.I. x D.E.:2.3×2.5"
- Nº de proveedor:284611000
This one-piece cylinder with handle provides a secure, solid grip for safer pouring.
- Polypropylene construction
- Secure, solid grip with handle
- Easy-to-read graduations
- Chemically resistant
- Autoclavable
These cylinders have an easy-to-read double scale with raised graduations, and a hexagonal base to prevent tipping and rolling. Made of polypropylene, it will not create a meniscus. Handle opening is 8.9 x 2.9 cm (3 x 1⅛"). Chemically resistant and autoclavable at 121ºC (250ºF).