PYREX® Lifetime Red™ Graduated Cylinders, To Contain, Double Pourout, Corning
Catalog # 89090-610
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Intervalo de graduación:2 mL
- Capacidad:100 ml
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:I-RECs, ISO 14001 - Monterrey
- Forma:Tall
- Referencia:89090-610
- Nº de proveedor:3044-100
Specifications
About this item
Borosilicate glass.
- Calibrated to contain (TC)
- Hexagonal base with graduations in white enamel
- Tuttle type of low form for stability with double pourout