Orders LinkContactUsLinkComponent
PYREXPLUS® Graduated Cylinders, To Deliver, Corning
PYREXPLUS® Graduated Cylinders, To Deliver, Corning
   Opciones sostenibles disponibles
Supplier:   Corning
PYREXPLUS® Graduated Cylinders, To Deliver, Corning
Supplier:  Corning
   Opciones sostenibles disponibles
Certificates
PYREXPLUS® Graduated Cylinders, To Deliver, Corning
PYREXPLUS® Graduated Cylinders, To Deliver, Corning
2 Options Available Below
Product Details & Documents
Borosilicate glass.
  • Calibrated to deliver (TD)
  • Graduations and markings in durable white enamel
  • Resistant to thermal shock
Cylinder features a protective polymer coating for longer product life and safety.
Coating makes cleanup faster and easier by reducing spill volume if breakage does occur. Coating is autoclavable to 121°C (249.8°F). Do not heat above 121°C (249.8°F) moist heat or 110°C (230°F) dry heat. Do not place coated ware over direct heat or open flames. Can be written on with any standard lab marker, permanent or water-soluble. Cylinder features single metric scale. Bumper guards not included.
PYREXPLUS® labware allows for a better hand grip and clamp hold.
Documents
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 89090-578
  • 89090-586
  • Form:
  • Tall
  • Tall
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Especificaciones
Product Family Options
Información del producto
  • Capacity
    Graduation Interval
    Tolerance
    Availability
    Price
  • 100 ml
    1 ml
    ±1.0 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 1
    Paquete de 1 - $1,910.00
    $1,910.00
    Log in Para disponibilidad
    Case de 4 (1UN)
    Case de 4 (1UN) - $4,338.26
    $4,338.26
    Paquete de 1
       $1,910.00
    Case de 4 (1UN)
       $4,338.26
    Supplier Number: 63024-100
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-578
    Form
    Tall
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2000 ml
    20 ml
    ±12.0 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 1
    Paquete de 1 - $7,513.87
    $7,513.87
    Log in Para disponibilidad
    Case de 2 (1UN)
    Case de 2 (1UN) - $15,027.72
    $15,027.72
    Paquete de 1
       $7,513.87
    Case de 2 (1UN)
       $15,027.72
    Supplier Number: 63024-2L
    Especificaciones:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-586
    Form
    Tall
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact