PYREX® Graduated Cylinders, To Contain, Corning
Proveedor: Corning
Certificados
Acerca de este artículo
Borosilicate glass.
- Calibrated to contain (TC In ) at 20 °C (68 °F)
- Hexagonal base with graduations in white enamel
- 10 ml cylinder has a funnel top
Use the 100 ml cylinder for ASTM E133, E1272, D86, D216, and D447.
Entrega: Supplied with bumper guards for cylinders of capacity 25 to 500 ml.
Especificaciones
- Catalog No:
- 24710-227
- 89090-568
- 89090-564
- 89090-560
- 24710-205
- 89090-552
- 89090-594
- Forma:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey