Nalgene® Square Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Designed for long-term storage of chemicals and specimens where sterility is not required.
- Leakproof
- Not autoclavable
Bottles provide excellent chemical resistance, and withstand temperatures from –100 to 120 °C (–148 to 248 °F). The square design is easy to handle, and saves space on benchtop or shelf, in refrigerators and freezers, and under the lab hood. Permanent graduations on all sizes.
Ordering information: Supplied with polypropylene screw caps assembled on bottles. For replacement caps for 16120-715 through -728, see 16219-122 series.
Specifications
- Catalog No:
- 16120-708
- 16120-715
- 16120-728
- 16120-719
- Environmentally Preferable:
- Product Transparency and Disclosure
- Product Transparency and Disclosure
- Product Transparency and Disclosure
- Product Transparency and Disclosure
- Certificación ambiental:
- ACT Label
- ACT Label
- ACT Label
- ACT Label