VWR® Hirsch Filter Funnels
Supplier: VWR International
Jump to:
About this item
These chemical-porcelain funnels feature a fixed perforated plate and glazing inside and out, except for rim.
- Highly resistant to chemicals
- Autoclavable
Filter funnels are available in a range of volume capacities and top outside diameters.
2 Options Available Below
Product Details & Documents
These chemical-porcelain funnels feature a fixed perforated plate and glazing inside and out, except for rim.
- Highly resistant to chemicals
- Autoclavable
Filter funnels are available in a range of volume capacities and top outside diameters.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- CapacityFits Paper DiameterTop O.D.AvailabilityPrice
- 10 ml10 mm42 mmEach (1UN)$182.18Case de 12 (1UN)$198.58Paleta de 3,360 (1UN)$612,133.54
- 20 ml13–15 mm55 mmEach (1UN)$217.15Case de 12 (1UN)$236.69