VWR® Buchner Funnels, Table-Type
750 ml, 109 mm
Catalog # 89038-112
Supplier: VWR International
Jump to:
Product Details & Documents
These chemical-porcelain funnels are ideal for vacuum filtration and feature a slanting bottom that leads to a tubulated side outlet.
- Perforated plate
- Autoclavable
Designed with a wide mouth for convenience and a wide base that rests firmly on a tabletop for stability.
Especificaciones
- Capacity:750 ml
- Height:109 mm