VWR® Laboratory Bottles, LDPE
Catalog # 414004-174
Supplier: VWR International
Specifications
- Tamaño de la tapa:28-410
- Capacidad:500 ml (16.9 oz.)
- Descripción:Narrow mouth bottles
- Referencia:414004-174
Specifications
About this item
Translucent, flexible bottles are for general laboratory use, and are especially good for shipping liquids and for trace metal analysis.
- Leakproof
- Translucent siding
Can be stored in freezer to −100 °C (−148 °F). Offer better visibility of contents than high-density polyethylene bottles, and have excellent chemical resistance to most acids, bases, and alcohols.
Delivery: Polypropylene screw closures included.