Graduated Addition Funnel, Fluoropolymer Resin Stopcock, Chemglass
80070-640
Small Business Enterprise
- [ST] unión No.:24/40
- Capacidad:250 ml
- Referencia:80070-640
- Nº de proveedor:CG-1712-03
Cylindrical style addition funnel graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line
With a [ST] outer joint and lower inner joint, both the same size, and pressure equalizing arm.