VITLAB® Reagent Bottles, with PP Screw Caps, BrandTech
83008-530
- Tamaño de la tapa:GL45
- Capacidad:5 L (1.3 gal.)
- Dimensiones:178 Dia. x 315 H, mm
- Referencia:83008-530
- Nº de proveedor:V100889
Slope-shouldered polypropylene bottles feature polypropylene screw caps.
- Bottles and caps are autoclavable
They provide excellent chemical resistance to aliphatic alcohols, aldehydes, aliphatic hydrocarbons, alkalis, and acids.
: For replacement caps, see 83009-032 series.
: 10 liter size includes two handles, five liter size includes a handle to facilitate movement.