Orders LinkContactUsLinkComponent
VWR® Disposable Funnels
VWR® Disposable Funnels
VWR® Disposable Funnels
Supplier:  VWR International
Certificates
VWR® Disposable Funnels
VWR® Disposable Funnels
2 Options Available Below
Product Details & Documents
Funnels are semi-transparent and are provided in a variety of stem lengths for varied pouring purposes.
  • Tapered at a 60° angle
  • Inside fluting
  • Polystyrene or polypropylene construction
Disposable funnels are inexpensive and can be purchased in bulk packs.
Documents
Product Family Options
Información del producto
  • Material
    Overall Height
    Stem Length
    Stem O.D.
    Top I.D.
    Availability
    Price
  • Polystyrene
    71.5 mm
    27 mm
    7.4 mm
    55 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $426.74
    $426.74
    Log in Para disponibilidad
    Paleta de 84 (100UN)
    Paleta de 84 (100UN) - $465.14
    $465.14
    Case (100UN)
       $426.74
    Paleta de 84 (100UN)
       $465.14
  • PP
    75 mm
    22.6 mm
    7.5 mm
    65 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $433.73
    $433.73
    Log in Para disponibilidad
    Paleta de 84 (100UN)
    Paleta de 84 (100UN) - $472.77
    $472.77
    Case (100UN)
       $433.73
    Paleta de 84 (100UN)
       $472.77