VWR® Analytical Funnels
Polypropylene analytical funnels feature a 60° body angle and 58° internal rib angle for rapid filtration.
- External ribbing prevents air lock
- Autoclavable
- Polypropylene fabrication
Resistant to most chemicals, except strong oxidizing agents.
Polypropylene analytical funnels feature a 60° body angle and 58° internal rib angle for rapid filtration.
- External ribbing prevents air lock
- Autoclavable
- Polypropylene fabrication
Resistant to most chemicals, except strong oxidizing agents.
Documents
- Catalog No:
- 414004-273
- 414004-274
- 414004-275
- 414004-276
- 414004-277
- 414004-278
- 414004-279
- 414004-280
- Temperature range:
- 0 to 135°C (32 to 275°F)
- 0 to 135°C (32 to 275°F)
- 0 to 135°C (32 to 275°F)
- 0 to 135°C (32 to 275°F)
- 0 to 135°C (32 to 275°F)
- 0 to 135°C (32 to 275°F)
- 0 to 135°C (32 to 275°F)
- 0 to 135°C (32 to 275°F)
Product Family Options
- Overall HeightStem LengthStem O.D.Top I.D.
- 76 mm49 mm6.4 mm35 mmPaquete de 12$77.91Case de 3 (12UN)$84.93Paleta de 1,386 (12UN)$107,987.70
Specifications
Cat. No.414004-273Temperature range0 to 135°C (32 to 275°F) - 85 mm49 mm7.2 mm45 mmPaquete de 12$84.39Case de 3 (12UN)$91.98Paleta de 300 (12UN)$25,316.34
Specifications
Cat. No.414004-274Temperature range0 to 135°C (32 to 275°F) - 105 mm60 mm7.2 mm55 mmPaquete de 12$90.82Case de 3 (12UN)$98.99Paleta de 300 (12UN)$27,245.64
Specifications
Cat. No.414004-275Temperature range0 to 135°C (32 to 275°F) - 114 mm62 mm6.5 mm65 mmPaquete de 12$103.20Case de 3 (12UN)$112.49Paleta de 180 (12UN)$18,576.22
Specifications
Cat. No.414004-276Temperature range0 to 135°C (32 to 275°F) - 141 mm77 mm6.9 mm75 mmPaquete de 6$65.03Case de 6 (6UN)$70.88Paleta de 180 (6UN)$11,705.29
Specifications
Cat. No.414004-277Temperature range0 to 135°C (32 to 275°F) - 164 mm89 mm8.2 mm90 mmPaquete de 6$136.51Case de 4 (6UN)$148.80Paleta de 300 (6UN)$40,953.48
Specifications
Cat. No.414004-278Temperature range0 to 135°C (32 to 275°F) - 180 mm96 mm9 mm100 mmPaquete de 4$109.65Case de 6 (4UN)$119.52Paleta de 180 (4UN)$19,737.72
Specifications
Cat. No.414004-279Temperature range0 to 135°C (32 to 275°F) - 278 mm154 mm14 mm160 mmPaquete de 2$109.65Case de 6 (2UN)$119.52Paleta de 108 (2UN)$11,842.63
Specifications
Cat. No.414004-280Temperature range0 to 135°C (32 to 275°F)