Nalgene® Laboratory Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Leakproof, translucent, flexible bottles, with excellent impact resistance and better visibility to contents than HDPE bottles.
- Excellent chemical resistance to most acids, bases, and alcohols
- LDPE is extremely low in trace metal content
- Excellent material for trace metal analysis
Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
: For replacement closures for 16058-112 and 16058-145, see 16219-122 series.
- Catalog No:
- 16057-980
- 16058-145
- 16058-043
- 16058-065
- 16058-000
- 16058-101
- 16058-123
- Environmentally Preferable:
- Low Manufacturing Impact
- Certificación ambiental:
- ISO14001 (Certificate Registration No. EMS 752603)
Frequently Bought Together