Neoprene Glove Box Gloves, Honeywell Safety
Proveedor: HONEYWELL SAFETY PRODUCTS USA INC.
Certificados
Acerca de este artículo
Premium quality milled neoprene gloves offer high tensile strength and flexibility.
- Seamless, one-piece construction provides maximum strength and integrity
- Ideal for applications requiring good sensitivity and dexterity
- Lead-loaded gloves contain 0.1 mm lead equivalent for attenuation of soft gamma radiation
Información de pedido: Butyl, Hypalon®, Neoprene/Hypalon® blend, and Viton® glove box gloves are also available (see 82031-206 series).
Especificaciones
- Catalog No:
- 32888-350
- Ambidiestro/Específico para cada mano:
- Específico para cada mano
Especificaciones
Información relacionada
Focus Safety Volume 1 Issue 1 Safety Essentials Volume 2
Documentación del producto
Frequently Bought Together