Orders LinkContactUsLinkComponent
Cryo-Gloves® Low-Temperature Gloves, Tempshield
Supplier:   TEMPSHIELD INC
Cryo-Gloves® Low-Temperature Gloves, Tempshield
Supplier:  TEMPSHIELD INC
Certificates
Cryo-Gloves® Low-Temperature Gloves, Tempshield
9 Options Available Below
Product Details & Documents
Protect hands and arms against ultracold temperatures to –196 °C (–321 °F).
  • Lightweight, breathable, comfortable
  • Provide warmth while retaining flexibility, even after prolonged exposure to cryogenic atmospheres
  • Offer superior thermal protection when working with ultra-low temperature freezers, blast freezers, dry ice, and in coldrooms
  • Also for use with LNG (liquefied natural gas), liquid helium, and liquid oxygen in enclosed systems
Gloves allow sufficient dexterity to handle small objects and controls. Waterproof gloves should be selected to protect the user from cryogenic liquids and vapors.
 : Gloves are not for immersion in LN₂.
Specifications for Product Family
Specifications
  • Catalog No:
  • 89166-343
  • 89166-359
  • 89426-964
  • 32885-758
  • 32884-072
  • 32884-074
  • 32884-085
  • 32884-021
  • 32884-031
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
Specifications
Product Family Options
Low-temperature glove
Product Information
  • Glove size
    Glove Color
    Length
    Availability
    Price
  • 89166-343
    Wrist length
    Wrist length
    M
    Pink
    29 - 32.5 cm (11⅜ - 12 1³⁄₁₆")
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $4,486.35
    $4,486.35
    Log in for availability
    Case de 10 (1Par)
    Case de 10 (1Par) - $4,890.12
    $4,890.12
    Par
       $4,486.35
    Case de 10 (1Par)
       $4,890.12
    Supplier Number: P-WRM
    Specifications:
    More
    Specifications
    Cat. No.
    89166-343
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
  • 89166-359
    Mid-arm length
    Mid-arm length
    M
    Pink
    34.5 - 39 cm (13⅝ - 15 ⁵⁄₁₆")
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $4,806.83
    $4,806.83
    Log in for availability
    Case de 10 (2UN)
    Case de 10 (2UN) - $5,239.43
    $5,239.43
    Par
       $4,806.83
    Case de 10 (2UN)
       $5,239.43
    Supplier Number: P-MAM
    Specifications:
    More
    Specifications
    Cat. No.
    89166-359
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
  • 89426-964
    Wrist length
    Wrist length
    L
    Blue
    29 - 32.5 cm (11⅜ - 12 1³⁄₁₆")
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $4,486.35
    $4,486.35
    Log in for availability
    Case de 10 (1Par)
    Case de 10 (1Par) - $4,890.12
    $4,890.12
    Par
       $4,486.35
    Case de 10 (1Par)
       $4,890.12
    Supplier Number: WRL
    Specifications:
    More
    Specifications
    Cat. No.
    89426-964
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
  • 32885-758
    Mid-arm length
    Mid-arm length
    L
    Blue
    34.5 - 39 cm (13⅝ - 15 ⁵⁄₁₆")
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $4,806.83
    $4,806.83
    Log in for availability
    Case de 10 (2UN)
    Case de 10 (2UN) - $5,239.43
    $5,239.43
    Par
       $4,806.83
    Case de 10 (2UN)
       $5,239.43
    Supplier Number: MAL
    Specifications:
    More
    Specifications
    Cat. No.
    32885-758
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
  • 32884-072
    Elbow length
    Elbow length
    S
    Blue
    44 - 48.3 cm (17⁵⁄₁₆ - 19")
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $5,982.91
    $5,982.91
    Log in for availability
    Case de 10 (1Par)
    Case de 10 (1Par) - $6,521.36
    $6,521.36
    Par
       $5,982.91
    Case de 10 (1Par)
       $6,521.36
    Supplier Number: EBS
    Specifications:
    More
    Specifications
    Cat. No.
    32884-072
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
  • 32884-074
    Elbow length
    Elbow length
    M
    Blue
    44 - 48.3 cm (17⁵⁄₁₆ - 19")
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $5,982.91
    $5,982.91
    Log in for availability
    Case de 10 (2UN)
    Case de 10 (2UN) - $6,521.36
    $6,521.36
    Par
       $5,982.91
    Case de 10 (2UN)
       $6,521.36
    Supplier Number: EBM
    Specifications:
    More
    Specifications
    Cat. No.
    32884-074
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
  • 32884-085
    Elbow length
    Elbow length
    L
    Blue
    44 - 48.3 cm (17⁵⁄₁₆ - 19")
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $5,982.91
    $5,982.91
    Log in for availability
    Case de 10 (2UN)
    Case de 10 (2UN) - $6,521.36
    $6,521.36
    Par
       $5,982.91
    Case de 10 (2UN)
       $6,521.36
    Supplier Number: EBL
    Specifications:
    More
    Specifications
    Cat. No.
    32884-085
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
Waterproof glove
Product Information
  • Glove size
    Glove Color
    Length
    Availability
    Price
  • 32884-021
    Wrist length
    Wrist length
    S
    Blue
    29 - 32.5 cm (11⅜ - 12 1³⁄₁₆")
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $5,501.66
    $5,501.66
    Log in for availability
    Case de 10 (1Par)
    Case de 10 (1Par) - $5,996.81
    $5,996.81
    Par
       $5,501.66
    Case de 10 (1Par)
       $5,996.81
    Supplier Number: WRSWP
    Specifications:
    More
    Specifications
    Cat. No.
    32884-021
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
  • 32884-031
    Mid-arm length
    Mid-arm length
    L
    Blue
    34.5 - 39 cm (13⅝ - 15 ⁵⁄₁₆")
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $5,928.94
    $5,928.94
    Log in for availability
    Case de 10 (1Par)
    Case de 10 (1Par) - $6,462.54
    $6,462.54
    Par
       $5,928.94
    Case de 10 (1Par)
       $6,462.54
    Supplier Number: MALWP
    Specifications:
    More
    Specifications
    Cat. No.
    32884-031
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable