Orders LinkContactUsLinkComponent
Trigger Spray Bottles, Qorpak
Trigger Spray Bottles, Qorpak
Supplier:   QORPAK
Trigger Spray Bottles, Qorpak
Supplier:  QORPAK
Certificates
Trigger Spray Bottles, Qorpak
Trigger Spray Bottles, Qorpak
1 Options Available Below
Product Details & Documents
Trigger spray bottles feature a trigger-motion spray that dispenses a spray or stream of liquid from an adjustable nozzle.
The translucent HDPE bottles are ideal for general use. HDPE provides an excellent moisture barrier and impact resistance. The adjustable sprayer permits a versatile spray pattern, from fine mist to coarse spray to jet stream.
Product Family Options
Información del producto
  • Capacity
    Material
    Cap Size
    Cap Type
    Availability
    Price
  • 16344-101
    Cylindrical
    Cylindrical
    473 ml (16 oz.)
    High-Density Polyethylene
    28-400
    White PP spray/stream trigger sprayer
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $107.31
    $107.31
    Log in Para disponibilidad
    Case de 6 (1UN)
    Case de 6 (1UN) - $643.89
    $643.89
    Each (1UN)
       $107.31
    Case de 6 (1UN)
       $643.89
    Supplier Number: PLC-03431
  • Oval
    650 ml (22 oz.)
    High-Density Polyethylene
    28-400
    White PP spray/stream trigger sprayer
    Discontinued
    Supplier Number: PLC-03499