ContactUsLinkComponent
Chemware® Wash Bottles, Teflon® PFA, Wide Mouth, Saint-Gobain
Catalog # 16651-880
undefined
Chemware® Wash Bottles, Teflon® PFA, Wide Mouth, Saint-Gobain
Catalog # 16651-880
Supplier Number:  D1069739
Specifications
  • Tamaño de la tapa:
    40 mm
  • Capacidad:
    100 ml (3.4 oz.)
  • Referencia:
    16651-880
  • Nº de proveedor:
    D1069739
Specifications
About this item
Transparent and unbreakable bottles can be rinsed with or used to dispense virtually all chemicals.
PFA fluoropolymer material withstands temperatures to 500 °F. Bottles are chemically inert, non-stick and easy to clean. Flex life and mechanical strength is superior to FEP. PFA contains the lowest concentration of trace metal ions of all fluoropolymers.