ContactUsLinkComponent
Bitran® Liquid-Tight Specimen Bags, Com-Pac
Bitran® Liquid-Tight Specimen Bags, Com-Pac
  11217-124
undefined
Bitran® Liquid-Tight Specimen Bags, Com-Pac
  11217-124
 :  4754-PE
  • Referencia:
    11217-124
  • An x La:
    180×200 mm (7×8")
  • Nº de proveedor:
    4754-PE
Polyethylene and SARANEX® zipper bags are designed for the handling and storage of specimens.
  • Unique manufacturing process eliminates leaks
  • Double track airtight zipper is easy to close
The SARANEX® (Series S) bags provide an exceptional vapor barrier for optimum odor control. Polyethylene (Series PE) bags remain flexible when frozen during storage.