SP Bel-Art Clavies™ Biohazard Autoclave Gloves, Bel-Art Products, a part of SP
Proveedor: Bel-Art Products, a Part of SP
Certificados
Acerca de este artículo
Soft, pliant, all-cotton terry cloth gloves are superb for handling hot objects out of the autoclave or oven, or for other heat protection in the lab, school or industry.
- Heat-resistant
- Machine wash, air dry
Heat-resistant up to 232 °C (450 °F), they replace hazardous asbestos gloves. Gauntlets in two lengths provide wrist and forearm protection, and the bright orange color offers high visibility.
Precaución: Gloves are not waterproof and should not be used to handle dry ice.
Información relacionada
Frequently Bought Together