Orders LinkContactUsLinkComponent
VWR® Water Sampling Kits
VWR® Water Sampling Kits
VWR® Water Sampling Kits
Supplier:  VWR International
Certificates
VWR® Water Sampling Kits
VWR® Water Sampling Kits
2 Options Available Below
Product Details & Documents
These sterile sampling bags are ideal for collecting chlorinated water specimens.
  • Sterile polyethylene construction
  • Suited for water sampling
  • Easy to use
  • Wire closure provides an airtight seal
They are made of sterile virgin polyethylene tubing with no side seals.
Bags feature a wire closure for an airtight seal and wide-sealed bottoms for added leakproof protection. They are easy to use: simply tear off perforated top, pull tabs to open bag, fill, grab ends of tape closure and pull shut, twirl bag three or four times to seal, and bend ends of closure inward to lock the seal.
UPA-approved for drinking water analysis.
Bags include a nontoxic, nutritive tablet containing 10 mg of active sodium thiosulfate.
Product Family Options
Información del producto
  • Capacity
    Thickness
    W×L
    Availability
    Price
  • 180 ml (6 oz.)
    2.5 mil.
    76×178 mm (3×7")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $1,773.30
    $1,773.30
    Case (100UN)
       $1,773.30
  • 540 ml (18 oz.)
    3 mil.
    114×229 mm (4.5×9")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (100UN)
    Case (100UN) - $2,166.16
    $2,166.16
    Case (100UN)
       $2,166.16