Orders LinkContactUsLinkComponent
VWR® Sample Sak™ Sample Bags
VWR® Sample Sak™ Sample Bags
12.7×7.6×22.8 cm (5×3×9")
Catalog # 11216-002
Supplier:   VWR International
VWR® Sample Sak™ Sample Bags
Catalog # 11216-002
Supplier:  VWR International
VWR® Sample Sak™ Sample Bags
VWR® Sample Sak™ Sample Bags
12.7×7.6×22.8 cm (5×3×9")
Price
$1,612.15
Log in Para disponibilidad
Paquete de 100 Case de 5 (100UN)
Product Details & Documents
Ideal collection/transport containers for powder and granular samples including grain, seeds, feeds, coffee, powdered soaps, and even damp soil.
  • For solid and semi-solid samples
  • Imprinted writing area
  • Tin tie tabs
Heat-sealed bags are constructed of 50 lb. machine finished white Kraft paper with tin tie tabs. Lined with 1.25 mm polyethylene. Imprinted on each bag is a personalized area for recording sample data and date.
Documents
Especificaciones
  • W×D×H:
    12.7×7.6×22.8 cm (5×3×9")