Butyl gloves provide the highest impermeability to water vapor, gases, and many toxic chemicals, including most missile fuels.
- Wide variety of styles and sizes to meet diverse glove box applications
Hypalon® gloves offer outstanding abrasion and chemical resistance, including resistance to ozone and oxidizing chemicals. Hypalon® is naturally soft for greater comfort.
Lead-loaded gloves contain 0.1 mm lead equivalent for attenuation of soft gamma radiation.
Neoprene/Hypalon® blend gloves combine the flexibility and dexterity of neoprene with the ozone and chemical resistance of Hypalon®. Extra long life and positive protection under unusually severe conditions. Excellent resistance to ozone and oxidizing chemicals. White surface for good visibility; will not hide contamination.
Viton® gloves are designed for handling chlorinated and aromatic solvents. Provide superior resistance to PCBs. Excellent permeation resistance to many chlorinated, aliphatic, and aromatic hydrocarbons, and most oils and acids. Highly flame retardant. Excellent dexterity.
Flexible and sensitive, even at low temperatures. Resistant to oxygenated solvents and most oxidizing chemicals.
- Catalog No:
- 56223-192
- 56223-160
- 11000-572
- Ambidiestro/Específico para cada mano:
- Específico para cada mano
- Específico para cada mano
- Específico para cada mano