Self-Locking Nylon Ties, Associated Bag
Supplier: ASSOCIATED BAG INC.
Jump to:
About this item
Tight locking to create a tamper-evident seal that must be cut to remove.
- Use to close bags, attach tags, bundle wires, make temporary repairs, and more
Higher tensile strengths for more demanding applications.
6 Options Available Below
Product Details & Documents
Tight locking to create a tamper-evident seal that must be cut to remove.
- Use to close bags, attach tags, bundle wires, make temporary repairs, and more
Higher tensile strengths for more demanding applications.
Documents
Product Family Options
Product Information
- ColorLengthWidthThicknessTensile StrengthAvailabilityPrice
- 89008-894Self-locking nylon tiesSupplier Number: 84-1-01R
- 80094-534Self-locking nylon tiesSelf-locking nylon tiesNatural5.5"0.13"0.048"40 lbs.Log in for availabilityPaquete de 1,000$1,056.76Supplier Number: 84-1-02
- 47744-096Self-locking nylon tiesSelf-locking nylon tiesGreen5.5"0.13"0.048"40 lbs.Log in for availabilityPaquete de 1,000$1,127.87Supplier Number: 84-1-02G
- 47745-022Self-locking nylon tiesSupplier Number: 84-1-03
- 47744-094Self-locking nylon tiesSupplier Number: 84-1-03G
- 10027-352Self-locking nylon tiesSupplier Number: 84-1-03L