PYREX® Snap Caps, Polyethylene, Corning
500
Supplier: Corning
Supplier #: 7666-500
PYREX® Snap Caps, Polyethylene, Corning
Catalog # 29610-579
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
These blue caps provide an economical closure for certain volumetric flasks (see 29610-004 and -285 series).
Documents
Especificaciones
- Cap Size:500
- Cat. No.:29610-579
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact