KIMBLE® RAY-SORB® Amber Heavy Duty Wide Mouth Volumetric Flask, Polyethylene Stopper, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
Flasks are made of red-tinted glass that provides protection from UV light
All flasks are graduated to Class A volumetric tolerances. Flasks with 5–250mL capacity are calibrated to meet ASTM Specification E288.
Feature wide mouth for easy pipet access and heavy, uniform walls for strength, durability, and safety. Interior bottoms are flat, allowing for efficient stirring with standard stir bars. Large, permanent markings allow for quick, accurate identification.
Related Information
Frequently Bought Together