ContactUsLinkComponent
KIMAX® Volumetric Flasks with [ST] Polyethylene Stopper, Wide Mouth, Class A, Kimble Chase
  89079-200
undefined
KIMAX® Volumetric Flasks with [ST] Polyethylene Stopper, Wide Mouth, Class A, Kimble Chase
  89079-200
 :  92812P-20
  • Capacidad:
    20 ml
  • Altura:
    135 mm
  • Referencia:
    89079-200
  • [ST] No. de tapón:
    13
  • Tolerancia:
    0.08 ml
  • Nº de proveedor:
    92812P-20
Flasks feature wide mouth for easy pipet access and heavy, uniform walls for strength, durability, and safety
All flasks are graduated to Class A volumetric tolerances. Flasks with 5–250mL capacity are calibrated to meet ASTM Specification E288.
Interior bottoms are flat, allowing for efficient stirring with standard stir bars. Large, permanent markings allow for quick, accurate identification.