Volumetric Flasks with [ST] Glass Stopper, Red Stripe, Class A, Hirschmann
Supplier: HIRSCHMANN INC
Certificates
About this item
Flasks feature a red stripe on neck for high contrast and easy meniscus reading, a fine white graduation line, and marking spot
Meet ASTM E288, Class A requirements.
Graduated to contain. Flasks include a [ST] glass stopper.
Frequently Bought Together