BCR® Polyester Glove Liners, Berkshire
32891-474
Small Business Enterprise
- Color del guante:White
- Tamaño del guante:Estándar
- Ambidiestro/Específico para cada mano:Ambidiestro
- Descripción:BCR® Polyester full-finger glove liner
- Referencia:32891-474
- Nº de proveedor:BGL3.20R
These medium weight glove liners are made out of high quality 100% contiuous filament polyester.
- Liners can be re-laundered and used again
- No seams on the gloves that could unravel and cause contamination
- Composition allows moisture to be absorbed and wicked away from skin during extended wear
- Ambidextrous for easy stock control
- Color coded cuffs to identify sizes
- Polyester glove liners can be sterilized by beta or steam
The half-finger versions of these liners are ideal for operators who need to retain maximum dexterity and fingertip sensitivity.
: Glove liners are sold by the pair.