Orders LinkContactUsLinkComponent
Nunc® CryoColor Coder Inserts, Thermo Scientific
Nunc® CryoColor Coder Inserts, Thermo Scientific
Certificates
Nunc® CryoColor Coder Inserts, Thermo Scientific
Nunc® CryoColor Coder Inserts, Thermo Scientific
5 Options Available Below
Product Details & Documents
For comprehensive sample identification, fits inside CryoTube® vial cap.
  • Made of polystyrene
  • Available in 10 different colors
 : Assortment 24270-120 includes either 50 of each color (for a pack) or 200 of each color (for a case).
Product Family Options
Product Information
  • Color
    Availability
    Price
  • Blue
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 500
    Paquete de 500 - $2,190.28
    $2,190.28
    Log in for availability
    Case de 4 (500UN)
    Case de 4 (500UN) - $2,387.40
    $2,387.40
    Paquete de 500
       $2,190.28
    Case de 4 (500UN)
       $2,387.40
    Supplier Number: 354879
  • Green
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 500
    Paquete de 500 - $2,190.28
    $2,190.28
    Log in for availability
    Case de 4 (500UN)
    Case de 4 (500UN) - $2,387.40
    $2,387.40
    Paquete de 500
       $2,190.28
    Case de 4 (500UN)
       $2,387.40
    Supplier Number: 355018
  • Purple
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 500
    Paquete de 500 - $2,155.19
    $2,155.19
    Log in for availability
    Case de 4 (500UN)
    Case de 4 (500UN) - $2,349.16
    $2,349.16
    Paquete de 500
       $2,155.19
    Case de 4 (500UN)
       $2,349.16
    Supplier Number: 375922
  • Red
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 500
    Paquete de 500 - $2,190.28
    $2,190.28
    Log in for availability
    Case de 4 (500UN)
    Case de 4 (500UN) - $2,387.40
    $2,387.40
    Paquete de 500
       $2,190.28
    Case de 4 (500UN)
       $2,387.40
    Supplier Number: 354968
  • Yellow
    Log in for availability
    Log in for availability
    Log in for availability
    Paquete de 500
    Paquete de 500 - $2,190.28
    $2,190.28
    Log in for availability
    Case de 4 (500UN)
    Case de 4 (500UN) - $2,387.40
    $2,387.40
    Paquete de 500
       $2,190.28
    Case de 4 (500UN)
       $2,387.40
    Supplier Number: 355077