Cotton Lisle Inspection Gloves, Wells Lamont®
Catalog # 32935-461
Supplier: WELLS LAMONT INDUSTRY GROUP
Specifications
- Color del guante:White
- Ambidiestro/Específico para cada mano:Ambidiestro
- Descripción:Lightweight
- Longitud:22.9 cm (9")
- Referencia:32935-461
- Nº de proveedor:Y6701W
Specifications
About this item
Hand and product protection for light-duty inspection purposes.
- Hand and product protection for light duty inspection purposes
- Lightweight and medium weight
- Men’s and women’s
- Cotton lisle (100%)
- Ambidextrous
The 100% cotton lisle inspection gloves come in lightweight and medium weight for men and women.
They are ambidextrous.