Orders LinkContactUsLinkComponent
VWR® Robo™ Thermometers
VWR® Robo™ Thermometers
VWR® Robo™ Thermometers
Supplier:  VWR International
Certificates
VWR® Robo™ Thermometers
2 Options Available Below
Product Details & Documents
Ideal for reading temperatures in reagents, water baths, foods, air, and semisolids.
  • Traceable®
  • Stainless steel probe
  • Rotating digital display
  • Read °C and °F
  • Extra-wide range
Probe is resistant to acids, bases, solvents, and most laboratory chemicals. Long stem allows user to make measurements while stirring solutions. Change between °F and °C with the press of a button. Readings are updated every second. Weigh less than 28g (1oz.).
Probe is resistant to acids, bases, solvents, and most laboratory chemicals. Long stem allows user to make measurements while stirring solutions. Change between °F and °C with the press of a button. Readings are updated every second. Weigh less than 28g (1 oz.).
Durable stainless steel and plastic construction makes these thermometers safer than mercury thermometers. Digital display conveniently rotates 180° to a vertical position, or to a horizontal position, for viewing sample and temperature simultaneously. Bright LCD digits are 6.4mm (1/4") high and easier to read than pointer or mercury thermometers.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a one-year warranty and 1.5V battery.
Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request.
Specifications for Product Family
Specifications
  • Catalog No:
  • 23609-204
  • 23609-206
  • Temperature range:
  • –50° to 280°C (–58° to 536°F)
  • –50° to 280°C (–58° to 536°F)
  • Probe:
  • 20.3L x 0.36 dia. cm (8 x 1/8")
  • 20.3L x 0.36 dia. cm (8 x 1/8")
Specifications
Product Family Options
Thermometer
Product Information
  • Accuracy
    Resolution
    Length
    Availability
    Price
  • 23609-204
    Robo™ Thermometer
    Robo™ Thermometer
    ±0.4 °C between –20 and 100 °C
    0.1° from –20° to 200°, 1° elsewhere
    23.5 cm (9¼")
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $2,538.98
    $2,538.98
    Each (1UN)
       $2,538.98
    Specifications:
    More
    Specifications
    Cat. No.
    23609-204
    Temperature range
    –50° to 280°C (–58° to 536°F)
    Probe
    20.3L x 0.36 dia. cm (8 x 1/8")
  • 23609-206
    Robo™ Thermometer
    Robo™ Thermometer
    ±1.0 °C between –20 and 100 °C
    0.1° from –20° to 200°, 1° elsewhere
    23.5 cm (9¼")
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,429.51
    $1,429.51
    Each (1UN)
       $1,429.51
    Specifications:
    More
    Specifications
    Cat. No.
    23609-206
    Temperature range
    –50° to 280°C (–58° to 536°F)
    Probe
    20.3L x 0.36 dia. cm (8 x 1/8")