ContactUsLinkComponent
Nalgene® Volumetric Flasks with Screw Cap, Polymethylpentene, Thermo Scientific
Nalgene® Volumetric Flasks with Screw Cap, Polymethylpentene, Thermo Scientific
Catalog # 29615-313
undefined
Nalgene® Volumetric Flasks with Screw Cap, Polymethylpentene, Thermo Scientific
Catalog # 29615-313
Supplier Number:  4001-0500
Specifications
  • Tamaño de la tapa:
    24
  • Capacidad:
    500 ml
  • Referencia:
    29615-313
  • Tolerancia:
    ±0.40 ml
  • Nº de proveedor:
    4001-0500
  • Altura con tapa:
    27.9 cm (11")
Specifications
About this item
Transparent polymethylpentene flasks are lighter in weight than glass and break resistant to protect against leakage and contamination
Each flask is individually hand calibrated to contain and to deliver at 20°C (68°F). Flasks contain a permanent color graduation line and oval marking spot for ease in reading fluid levels. Autoclavable, leakproof, and graduated.
Certifications: Flasks meet ASTM E288 and ISO 384 Class B tolerances for consistent quality and dependability.