PYREX® Volumetric Flasks, Phosphoric Acid, Wide Neck, Corning
Catálogo # 89090-794
Proveedor: Corning
Product available on GSA Advantage®
Especificaciones
- Capacidad:250 ml
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:I-RECs, ISO 14001 - Monterrey
- Referencia:89090-794
- Tolerancia:±0.18 ml
- Nº de proveedor:5840-250
Especificaciones
Acerca de este artículo
PYREX® borosilicate glass.
- Calibrated 'to contain'
- Heavily constructed throughout
For use in the determination of phosphoric acid in mixed fertilizer and more.